Gene Transcripts

Gene Symbol
RGD1561849
Entrez Gene ID
500393
Gene Name
similar to RIKEN cDNA 3110035E14
Gene Aliases
Chromosome Location
Chr. 5: 9355490 - 9381161 on Build Rnor_6.0
UniGene ID
Rn.18085
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109260.1 NP_001102730.1 1 412
GenBank mRNA FQ211566.1 - 1 326
FQ212089.1 - 1 336
FQ212229.1 - 1 336

View Full Product Details