Gene Transcripts

Gene Symbol
Rnf144a
Entrez Gene ID
500636
Gene Name
ring finger protein 144A
Gene Aliases
RGD1561737, Rnf144
Chromosome Location
Chr. 6: 45509895 - 45629161 on Build Rnor_6.0
UniGene ID
Rn.214624
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082410.1 NP_001075879.1 5 575
XM_008764604.1 XP_008762826.1 4 809

View Full Product Details