Gene Transcripts

Gene Symbol
Dtnbp1
Entrez Gene ID
641528
Gene Name
dystrobrevin binding protein 1
Gene Aliases
Chromosome Location
Chr. 17: 20090136 - 20182332 on Build Rnor_6.0
UniGene ID
Rn.187290
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037664.1 NP_001032753.1 9 951
GenBank mRNA AY623026.1 - 1525
BC088267.1 - 9 951 7379090

View Full Product Details