Gene Transcripts

Gene Symbol
Sf3b5
Entrez Gene ID
680891
Gene Name
splicing factor 3b, subunit 5
Gene Aliases
Chromosome Location
Chr. 1: 7101715 - 7102427 on Build Rnor_6.0
UniGene ID
Rn.2083
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126092.2 NP_001119564.1 1 275
GenBank mRNA CB725243.1 - 1 275

View Full Product Details