Gene Transcripts

Gene Symbol
GREB1
Entrez Gene ID
9687
Gene Name
growth regulation by estrogen in breast cancer 1
Gene Aliases
Chromosome Location
Chr. 2: 11534116 - 11642788 on Build GRCh38
UniGene ID
Hs.467733
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014668.3 NP_055483.2 5 884
NM_033090.2 NP_149081.1 5 835
NM_148903.2 NP_683701.2 5 900
XM_005246192.4 XP_005246249.1 5 906
XM_011510418.2 XP_011508720.1 4 745
XM_011510419.2 XP_011508721.1 4 745
XM_011510423.2 XP_011508725.1 4 745
GenBank mRNA AF245388.1 - 5 884
AF245389.1 - 5 835
AF245390.1 - 5 900
BC071853.1 - 4 565

View Full Product Details