Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008770390.1 | XP_008768612.1 | 27 | 4170 | |
XM_008770391.1 | XP_008768613.1 | 27 | 4149 | ||
XM_008770392.1 | XP_008768614.1 | 26 | 3996 | ||
XM_008770393.1 | XP_008768615.1 | 22 | 4912 |