Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191072.1 | NP_001178001.1 | 2, 3 | 437 | |
XM_008762383.1 | XP_008760605.1 | 2, 3 | 521 | ||
XM_008762384.1 | XP_008760606.1 | 2, 3 | 1312 | ||
XM_008762385.1 | XP_008760607.1 | 2, 3 | 1700 |