Gene Transcripts

Gene Symbol
KIAA0101
Entrez Gene ID
9768
Gene Name
KIAA0101
Gene Aliases
L5, NS5ATP9, OEATC, OEATC-1, OEATC1, PAF, PAF15, p15(PAF), p15/PAF, p15PAF
Chromosome Location
Chr. 15: 64364994 - 64387687 on Build GRCh38
UniGene ID
Hs.81892
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029989.2 NP_001025160.1 3 493
NM_014736.5 NP_055551.1 4 656
NR_109934.1 4 712
GenBank mRNA AK290748.1 - 4 587
AV757440.1 - 3 523
BC005832.2 - 4 576 2988995
BC007101.1 - 4 583 4281110
BC016782.1 - 4 591 4109322
D14657.1 - 4 578

View Full Product Details