Gene Transcripts

Gene Symbol
URB2
Entrez Gene ID
9816
Gene Name
URB2 ribosome biogenesis 2 homolog (S. cerevisiae)
Gene Aliases
KIAA0133, NET10, NPA2
Chromosome Location
Chr. 1: 229626216 - 229660200 on Build GRCh38
UniGene ID
Hs.533628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001314021.1 NP_001300950.1 3 405
NM_014777.3 NP_055592.2 3 374
XM_005273360.3 XP_005273417.1 3 343
GenBank mRNA AK291013.1 - 3 374
BC059381.1 - 3 349
BC114559.1 - 3 327 40082897
BC114963.1 - 3 296 40082900
D50923.2 - 3 356
DC369323.1 - 3 374

View Full Product Details