Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012296.3 | NP_036428.1 | 7 | 1634 | |
NM_080491.2 | NP_536739.1 | 7 | 1690 | ||
XM_006718753.2 | XP_006718816.1 | 8 | 1735 | ||
XM_011545408.2 | XP_011543710.1 | 6 | 1341 | ||
GenBank mRNA | AB011143.1 | - | 7 | 1634 | |
AB018413.1 | - | 7 | 1588 | ||
BC131711.1 | - | 7 | 1592 | 40122341 | |
BC152459.1 | - | 7 | 1634 | 8862595 |