Gene Transcripts

Gene Symbol
TBC1D4
Entrez Gene ID
9882
Gene Name
TBC1 domain family member 4
Gene Aliases
AS160, NIDDM5
Chromosome Location
Chr. 13: 75284664 - 75482646 on Build GRCh38
UniGene ID
Hs.210891
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286658.2 NP_001273587.1 20 4220
NM_001286659.2 NP_001273588.1 19 4055
NM_014832.4 NP_055647.2 21 4244
XM_005266603.1 XP_005266660.1 20 4647
XM_005266605.2 XP_005266662.1 21 3572
XM_006719903.3 XP_006719966.1 21 3932
XM_011535331.1 XP_011533633.1 20 4611
XM_017020882.1 XP_016876371.1 19 3264
XM_017020883.1 XP_016876372.1 18 3258
XM_017020884.1 XP_016876373.1 9 2830
GenBank mRNA AB011175.1 - 22 4193
AB449885.1 - 22 3846
AK300468.1 - 10 1753
AK304091.1 - 9 1574
BC151239.1 - 22 4185 8860512
FM207106.1 - 19 3654
FM207107.1 - 20 3819

View Full Product Details