Gene Transcripts

Gene Symbol
Pabpc5
Entrez Gene ID
688518
Gene Name
poly A binding protein, cytoplasmic 5
Gene Aliases
Chromosome Location
Chr. X: 92130292 - 92132442 on Build Rnor_6.0
UniGene ID
Rn.218841
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110366.2 NP_001103836.1 1 974
XM_008773376.1 XP_008771598.1 2 1365

View Full Product Details