Gene Transcripts

Gene Symbol
Mlkl
Entrez Gene ID
690743
Gene Name
mixed lineage kinase domain-like
Gene Aliases
Chromosome Location
Chr. 19: 43508416 - 43535051 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772570.1 XP_008770792.1 5 992
XM_008772571.1 XP_008770793.1 5 947
XM_008772572.1 XP_008770794.1 4 858

View Full Product Details