Gene Transcripts

Gene Symbol
FGF19
Entrez Gene ID
9965
Gene Name
fibroblast growth factor 19
Gene Aliases
Chromosome Location
Chr. 11: 69698238 - 69704338 on Build GRCh38
UniGene ID
Hs.249200
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005117.2 NP_005108.1 3 2018
GenBank mRNA AF110400.1 - 3 2018
AY358302.1 - 3 2018
BC017664.1 - 3 1686 3849343

View Full Product Details