Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001076793.1 | NP_001070261.1 | 3 | 178 | |
XM_006228225.2 | XP_006228287.1 | 2 | 399 | ||
XM_008758988.1 | XP_008757210.1 | 3 | 201 | ||
XM_008758989.1 | XP_008757211.1 | 2 | 165 | ||
XM_008758990.1 | XP_008757212.1 | 3 | 201 | ||
GenBank mRNA | AY998481.1 | - | 3 | 178 |