Gene Transcripts

Gene Symbol
FLT3LG
Entrez Gene ID
2323
Gene Name
fms related tyrosine kinase 3 ligand
Gene Aliases
FL, FLT3L
Chromosome Location
Chr. 19: 49474172 - 49487038 on Build GRCh38
UniGene ID
Hs.428
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204502.1 NP_001191431.1 9 943
NM_001204503.1 NP_001191432.1 8 912
NM_001278637.1 NP_001265566.1 9 920
NM_001278638.1 NP_001265567.1 9 909
NM_001459.3 NP_001450.2 9 892
XM_005258680.4 XP_005258737.3 8 828
XM_005258681.4 XP_005258738.3 8 879
XM_005258682.4 XP_005258739.3 7 795
XM_005258683.4 XP_005258740.3 7 743
XM_011526675.2 XP_011524977.1 8 749
XM_011526676.2 XP_011524978.1 8 714
XM_011526678.2 XP_011524980.1 8 695
XM_011526680.2 XP_011524982.1 5 495
XM_011526682.2 XP_011524984.1 8 899
XM_017026533.1 XP_016882022.1 7 639
XM_017026534.1 XP_016882023.1 7 612
XM_017026535.1 XP_016882024.1 9 861
GenBank mRNA BC006331.2 - 9 882
BC011914.2 - 9 882
BC028001.1 - 9 893 5198812
BC126293.1 - 9 849 8992009
BC136464.1 - 9 849 9020453
BC144129.1 - 9 900 9052649

View Full Product Details