Gene Transcripts

Gene Symbol
GLI4
Entrez Gene ID
2738
Gene Name
GLI family zinc finger 4
Gene Aliases
HKR4, ZNF928
Chromosome Location
Chr. 8: 143267437 - 143276931 on Build GRCh38
UniGene ID
Hs.400533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138465.3 NP_612474.1 4 1315
GenBank mRNA AY129020.1 - 1 2306
BC014165.2 - 3 1281 4549240

View Full Product Details