Gene Transcripts

Gene Symbol
GYPE
Entrez Gene ID
2996
Gene Name
glycophorin E (MNS blood group)
Gene Aliases
GPE, MNS, MiIX
Chromosome Location
Chr. 4: 143870866 - 143912291 on Build GRCh38
UniGene ID
Hs.654368
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002102.3 NP_002093.2 3 289
NM_198682.2 NP_941391.2 3 289
XM_017008139.1 XP_016863628.1 4 4216
XM_017008140.1 XP_016863629.1 4 4216
GenBank mRNA BC017864.1 - 3 285
BC064396.1 - 3 246
BG528203.1 - 3 285
BX091225.1 - 3 257
M29610.1 - 3 289

View Full Product Details