Gene Transcripts

Gene Symbol
INSL5
Entrez Gene ID
10022
Gene Name
insulin like 5
Gene Aliases
PRO182, UNQ156
Chromosome Location
Chr. 1: 66797741 - 66801259 on Build GRCh38
UniGene ID
Hs.251380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005478.4 NP_005469.2 2 243
GenBank mRNA AF133816.2 - 2 252
AY359030.1 - 2 243
BC101646.1 - 2 232 8069152
BC101648.1 - 2 232 8069154

View Full Product Details