Gene Transcripts

Gene Symbol
IFITM3
Entrez Gene ID
10410
Gene Name
interferon induced transmembrane protein 3
Gene Aliases
1-8U, DSPA2b, IP15
Chromosome Location
Chr. 11: 319673 - 320914 on Build GRCh38
UniGene ID
Hs.374650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021034.2 NP_066362.2 2 497
NR_049759.1 2 492
GenBank mRNA AK292173.1 - 2 441
BC006794.1 - 2 455 2986145
BC008417.1 - 2 440 4075453
BC022439.1 - 2 446 4282809
BC070243.1 - 2 431 30401041
BQ937265.1 - 2 300
DB261103.1 - 2 497
X57352.1 - 2 633

View Full Product Details