Gene Transcripts

Gene Symbol
TMEM52
Entrez Gene ID
339456
Gene Name
transmembrane protein 52
Gene Aliases
Chromosome Location
Chr. 1: 1917590 - 1919301 on Build GRCh38
UniGene ID
Hs.123423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178545.3 NP_848640.1 4, 5 379
GenBank mRNA AJ278736.1 - 4, 5 393
AY358544.1 - 1, 2 539
CN263898.1 - 4, 5 391

View Full Product Details