Gene Transcripts

Gene Symbol
WDR86
Entrez Gene ID
349136
Gene Name
WD repeat domain 86
Gene Aliases
Chromosome Location
Chr. 7: 151377974 - 151410705 on Build GRCh38
UniGene ID
Hs.647083
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284260.1 NP_001271189.1 5 1465
NM_001284261.1 NP_001271190.1 4 766
NM_001284262.1 NP_001271191.1 5 902
NM_198285.2 NP_938026.2 5 1401
XM_005249989.3 XP_005250046.1 4 1262
XM_005249990.4 XP_005250047.1 5 1398
XM_011516144.2 XP_011514446.1 5 1462
XM_011516145.2 XP_011514447.1 5 1462
XM_011516146.2 XP_011514448.1 5 1398
XM_011516147.2 XP_011514449.1 4 1262
XM_011516148.1 XP_011514450.1 5 950
XM_011516150.1 XP_011514452.1 6 1154
XM_011516151.2 XP_011514453.1 4 1262
GenBank mRNA AK125347.1 - 4 715
AK296049.1 - 5 1058
BC107125.2 - 4 798 40023951
BX390499.1 - 3 508

View Full Product Details