Gene Transcripts

Gene Symbol
KRTAP10-5
Entrez Gene ID
386680
Gene Name
keratin associated protein 10-5
Gene Aliases
KAP10.5, KAP18-5, KAP18.5, KRTAP10.5, KRTAP18-5, KRTAP18.1, KRTAP18.5
Chromosome Location
Chr. 21: 44579455 - 44580604 on Build GRCh38
UniGene ID
Hs.688629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198694.3 NP_941967.3 1 549
GenBank mRNA AB076352.1 - 1 549
BC120949.1 - 1 548 40115143
BC120950.1 - 1 548 40115146

View Full Product Details