Gene Transcripts

Gene Symbol
BGN
Entrez Gene ID
633
Gene Name
biglycan
Gene Aliases
DSPG1, PG-S1, PGI, SLRR1A
Chromosome Location
Chr. X: 153494889 - 153509554 on Build GRCh38
UniGene ID
Hs.821
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001711.5 NP_001702.1 6 941
XM_017029724.1 XP_016885213.1 5 720
GenBank mRNA AK092954.1 - 6 720
AK093023.1 - 6 775
AK094059.1 - 7 750
AK222484.1 - 6 850
AK223302.1 - 6 821
AK291955.1 - 6 852
AK293289.1 - 6 776
AK297966.1 - 6 637
BC002416.2 - 6 834 3162633
BC004244.1 - 6 844 3503374
BT007323.1 - 5 705
J04599.1 - 6 825

View Full Product Details