Gene Transcripts

Gene Symbol
ASAH2
Entrez Gene ID
56624
Gene Name
N-acylsphingosine amidohydrolase 2
Gene Aliases
BCDase, HNAC1, LCDase, N-CDase, NCDase
Chromosome Location
Chr. 10: 50184861 - 50251564 on Build GRCh38
UniGene ID
Hs.512645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143974.1 NP_001137446.1 19 2246
NM_019893.2 NP_063946.2 20 2351
XM_011539970.2 XP_011538272.1 21 2547
XM_011539971.2 XP_011538273.1 20 2404
XM_011539972.2 XP_011538274.1 20 2371
XM_017016435.1 XP_016871924.1 22 2518
XM_017016436.1 XP_016871925.1 6 610
GenBank mRNA BG289059.1 - 4 403

Gene Symbol
ASAH2B
Entrez Gene ID
653308
Gene Name
N-acylsphingosine amidohydrolase 2B
Gene Aliases
ASAH2C, ASAH2L, bA449O16.3, bA98I6.3
Chromosome Location
Chr. 10: 50739688 - 50757188 on Build GRCh38
UniGene ID
Hs.659820
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079516.3 NP_001072984.1 6 831
NM_001321957.1 NP_001308886.1 6 812
NM_001321958.1 NP_001308887.1 6 886
NM_001321959.1 NP_001308888.1 6 905
NM_001321960.1 NP_001308889.1 7 974
NR_135904.1 5 739
GenBank mRNA AK294369.1 - 6 638
BG773189.1 - 6 575
CN428611.1 - 5 481

View Full Product Details