Gene Transcripts

Gene Symbol
ATP9B
Entrez Gene ID
374868
Gene Name
ATPase phospholipid transporting 9B (putative)
Gene Aliases
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
Chromosome Location
Chr. 18: 79069275 - 79378391 on Build GRCh38
UniGene ID
Hs.465475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK130839.1 - 1 750

View Full Product Details