Gene Transcripts

Gene Symbol
ZDHHC11B
Entrez Gene ID
653082
Gene Name
zinc finger DHHC-type containing 11B
Gene Aliases
DHHC-11B
Chromosome Location
Chr. 5: 710355 - 784749 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017010113.1 XP_016865602.1 1 151
XM_017010115.1 XP_016865604.1 1 151

View Full Product Details