Gene Transcripts

Gene Symbol
DHRS4L1
Entrez Gene ID
728635
Gene Name
dehydrogenase/reductase 4 like 1
Gene Aliases
SDR25C4
Chromosome Location
Chr. 14: 24007009 - 24051377 on Build GRCh38
UniGene ID
Hs.647569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_102693.1 3 507
XM_006720246.3 XP_006720309.1 3 352
GenBank mRNA BC171918.1 - 3 466

View Full Product Details