Gene Transcripts

Gene Symbol
MPHOSPH9
Entrez Gene ID
10198
Gene Name
M-phase phosphoprotein 9
Gene Aliases
MPP-9, MPP9
Chromosome Location
Chr. 12: 123152324 - 123244014 on Build GRCh38
UniGene ID
Hs.577404
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022782.3 NP_073619.3 22 3488
NR_103517.1 22 3227
XM_011537740.2 XP_011536042.1 22 3392
XM_011537741.2 XP_011536043.1 22 3392
XM_017018673.1 XP_016874162.1 22 3486
XM_017018674.1 XP_016874163.1 21 3290
XM_017018675.1 XP_016874164.1 21 3294
XM_017018676.1 XP_016874165.1 21 3236
XM_017018677.1 XP_016874166.1 20 3167
XM_017018678.1 XP_016874167.1 21 3212
XM_017018679.1 XP_016874168.1 23 3425
XM_017018680.1 XP_016874169.1 21 3334
XM_017018681.1 XP_016874170.1 21 3247
GenBank mRNA AK091181.1 - 18 2817
AK096541.1 - 22 3100
AL096751.1 - 15 2044
BC130440.1 - 18 2770 40146471
BC144016.1 - 16 2540
BC144017.1 - 17 2422
X98258.1 - 4 270

View Full Product Details