Gene Transcripts
- Gene Symbol
- Unc5b
- Entrez Gene ID
- 107449
- Gene Name
- unc-5 homolog B (C. elegans)
- Gene Aliases
- 6330415E02Rik, A630020F16, D10Bwg0792e, Unc5h2
- Chromosome Location
- Chr. 10: 60762594 - 60831604 on Build GRCm38
- UniGene ID
- Mm.290433
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029770.2 |
NP_084046.2
|
5, 6 |
1143 |
|
GenBank mRNA |
AJ487853.1 |
- |
5, 6 |
1046 |
|
AK018177.1 |
- |
5, 6 |
1143 |
|
AK147603.1 |
- |
5, 6 |
1038 |
|
AK154271.1 |
- |
5, 6 |
1143 |
|
AK154287.1 |
- |
5, 6 |
1146 |
|
AK158284.1 |
- |
5, 6 |
1143 |
|
AK170513.1 |
- |
5, 6 |
1142 |
|
BC048162.1 |
- |
5, 6 |
950 |
|
BC057560.1 |
- |
5, 6 |
950 |
6417563 |
View Full Product Details