Gene Transcripts

Gene Symbol
Ssc4d
Entrez Gene ID
109267
Gene Name
scavenger receptor cysteine rich family, 4 domains
Gene Aliases
C330016E03Rik, S4D-SRCRB, SRCRB-S4D, Srcrb4d
Chromosome Location
Chr. 5: 135960220 - 135974535 on Build GRCm38
UniGene ID
Mm.332750
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160366.1 NP_001153838.1 11 1754
GenBank mRNA AK082786.1 - 3 165
BC117747.1 - 8 1261
BC126947.1 - 8 1261

View Full Product Details