Gene Transcripts

Gene Symbol
P2ry6
Entrez Gene ID
233571
Gene Name
pyrimidinergic receptor P2Y, G-protein coupled, 6
Gene Aliases
2010204J23Rik, P2Y6
Chromosome Location
Chr. 7: 100937630 - 100975492 on Build GRCm38
UniGene ID
Mm.235193
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183168.2 NP_898991.1 3 1600
GenBank mRNA AK142680.1 - 3 1625
BC027331.1 - 3 1596 3982042
BC064095.1 - 3 1580 1054566

View Full Product Details