Gene Transcripts

Gene Symbol
Olfr545
Entrez Gene ID
258837
Gene Name
olfactory receptor 545
Gene Aliases
MOR42-1, S50
Chromosome Location
Chr. 7: 102493823 - 102494773 on Build GRCm38
UniGene ID
Mm.377714
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146840.1 NP_667051.1 1 590
GenBank mRNA BC106987.1 - 1 590
BC128265.1 - 1 590 40043266
BC128266.1 - 1 590 40043268

View Full Product Details