Gene Transcripts

Gene Symbol
Ano8
Entrez Gene ID
382014
Gene Name
anoctamin 8
Gene Aliases
BC053460, Tmem16h
Chromosome Location
Chr. 8: 71476017 - 71486067 on Build GRCm38
UniGene ID
Mm.260360
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164679.1 NP_001158151.1 16 2756
GenBank mRNA AK173222.1 - 14 3744
BC027735.1 - 16 2658
BC027790.1 - 8 2324
BC042391.1 - 9 2951
BC053460.1 - 4 795
BC059855.1 - 7 1289

View Full Product Details