Gene Transcripts

Gene Symbol
Eif1
Entrez Gene ID
20918
Gene Name
eukaryotic translation initiation factor 1
Gene Aliases
Sui1-rs1
Chromosome Location
Chr. 11: 100319994 - 100322096 on Build GRCm38
UniGene ID
Mm.13886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011508.1 NP_035638.1 3 360
GenBank mRNA AF129888.1 - 3 345
AK148032.1 - 3 357
AK150813.1 - 3 359
AK151070.1 - 3 357
AK151575.1 - 3 359
AK152194.1 - 3 359
AK169763.1 - 3 358
BC003463.1 - 3 360 2648657
BC010791.1 - 3 338 4218816
BC081429.1 - 3 347 5710951
BC096656.1 - 3 335 5143414
BC096691.1 - 3 345 5256396
Z50159.1 - 2 103

View Full Product Details