Gene Transcripts

Gene Symbol
Mapk8ip2
Entrez Gene ID
60597
Gene Name
mitogen-activated protein kinase 8 interacting protein 2
Gene Aliases
3230402N03Rik, AI847694, AW049958, IB2, Jip2, R75148
Chromosome Location
Chr. 15: 89453911 - 89462447 on Build GRCm38
UniGene ID
Mm.173337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021921.3 NP_068740.3 5 927
GenBank mRNA AF220195.1 - 5 646
AF310135.1 - 5 708
AK014339.1 - 5 631
AK158968.1 - 5 781
BC029704.1 - 5 706 5359447

View Full Product Details