Gene Transcripts
- Gene Symbol
- Igkv1-135
- Entrez Gene ID
- 243420
- Gene Name
- immunoglobulin kappa variable 1-135
- Gene Aliases
- Gm4964, bd2
- Chromosome Location
- Chr. 6: 67609745 - 67610508 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
- Gene Symbol
- Igkc
- Entrez Gene ID
- 16071
- Gene Name
- immunoglobulin kappa constant
- Gene Aliases
- Igk-C
- Chromosome Location
- Chr. 6: 70726435 - 70726754 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC092090.1 |
- |
|
229 |
|
Z22077.1 |
- |
|
163 |
|
- Gene Symbol
- Igkv1-133
- Entrez Gene ID
- 628027
- Gene Name
- immunoglobulin kappa variable 1-133
- Gene Aliases
- EG628027, Gm6827, Igk-Vk2(70/1)
- Chromosome Location
- Chr. 6: 67724914 - 67725661 on Build GRCm38
- UniGene ID
- Mm.333124
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
Z17401.1 |
- |
2 |
235 |
|
View Full Product Details