Gene Transcripts

Gene Symbol
SLC30A9
Entrez Gene ID
10463
Gene Name
solute carrier family 30 member 9
Gene Aliases
C4orf1, GAC63, HUEL, ZNT9
Chromosome Location
Chr. 4: 41990472 - 42087534 on Build GRCh38
UniGene ID
Hs.479634
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006345.3 NP_006336.3 4, 5 585
XM_017007654.1 XP_016863143.1 4, 5 619
GenBank mRNA AF006621.2 - 4, 5 559
AK225470.1 - 4, 5 522
AK299917.1 - 5, 6 678
AK312838.1 - 4, 5 585
AY319413.1 - 5, 6 464
AY594282.1 - 4, 5 439
BC000240.1 - 4, 5 546
BC007732.2 - 4, 5 546 4109702
BC016949.1 - 4, 5 564 3850521
BG323207.1 - 586
KU178526.1 - 4, 5 439
KU178527.1 - 4, 5 439

View Full Product Details