Gene Transcripts

Gene Symbol
Sdhc
Entrez Gene ID
66052
Gene Name
succinate dehydrogenase complex, subunit C, integral membrane protein
Gene Aliases
0610010E03Rik, AI316496, AU019277
Chromosome Location
Chr. 1: 171129157 - 171150603 on Build GRCm38
UniGene ID
Mm.198138
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025321.3 NP_079597.2 3 125
GenBank mRNA AK002459.1 - 3 127
AK012818.1 - 3 126
AK032458.1 - 127
AK135503.1 - 3 127
BC005779.1 - 3 112 2810852
BY711932.1 - 3 126

View Full Product Details