Gene Transcripts

Gene Symbol
Qprt
Entrez Gene ID
67375
Gene Name
quinolinate phosphoribosyltransferase
Gene Aliases
2410027J01Rik, AI647766, QPRTase
Chromosome Location
Chr. 7: 127107770 - 127122029 on Build GRCm38
UniGene ID
Mm.26928
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133686.1 NP_598447.1 4 1037
GenBank mRNA AK010602.1 - 4 1051
AK019119.1 - 4 1052
BC011191.1 - 4 1037 4196244

View Full Product Details