Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026348.3 | NP_080624.1 | 6 | 418 | |
GenBank mRNA | AK005779.1 | - | 6 | 487 | |
AK013373.1 | - | 6 | 433 | ||
AK015557.1 | - | 6 | 443 | ||
AK045037.1 | - | 6 | 419 | ||
AK165072.1 | - | 4 | 274 | ||
AK166362.1 | - | 6 | 419 | ||
AK179730.1 | - | 4 | 224 | ||
BC049557.1 | - | 6 | 428 | 6741650 |