Gene Transcripts

Gene Symbol
Ooep
Entrez Gene ID
67968
Gene Name
oocyte expressed protein
Gene Aliases
2410146L05Rik, Floped, Moep19, Sddr
Chromosome Location
Chr. 9: 78376903 - 78378588 on Build GRCm38
UniGene ID
Mm.26883
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026480.3 NP_080756.1 2 248
GenBank mRNA AB283026.1 - 2 188
AK010266.1 - 2 243
AK010809.1 - 2 249
AK133554.1 - 2 213
AK135950.1 - 2 214
AK136199.1 - 214
AK145510.1 - 2 214
BC062245.1 - 2 199 3470921
DQ238107.1 - 2 249

View Full Product Details