Gene Transcripts
- Gene Symbol
- Mrps9
- Entrez Gene ID
- 69527
- Gene Name
- mitochondrial ribosomal protein S9
- Gene Aliases
- 2310002A08Rik, AW322516, Mrp-S9, S9mt, mg637
- Chromosome Location
- Chr. 1: 42851233 - 42905683 on Build GRCm38
- UniGene ID
- Mm.252982
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023514.4 |
NP_076003.3
|
3, 4 |
445 |
|
GenBank mRNA |
AK009082.1 |
- |
3, 4 |
395 |
|
AK031497.1 |
- |
3, 4 |
394 |
|
AK049465.1 |
- |
3, 4 |
403 |
|
AK144656.1 |
- |
3, 4 |
403 |
|
AK150985.1 |
- |
3, 4 |
403 |
|
BC051936.1 |
- |
2, 3 |
138 |
|
BC069980.1 |
- |
1, 2 |
39 |
|
BC083159.1 |
- |
2, 3 |
206 |
|
BC116437.2 |
- |
3, 4 |
395 |
40104493 |
View Full Product Details