Gene Transcripts

Gene Symbol
Cdk13
Entrez Gene ID
69562
Gene Name
cyclin-dependent kinase 13
Gene Aliases
2310015O17Rik, Cdc2l5
Chromosome Location
Chr. 13: 17715962 - 17805332 on Build GRCm38
UniGene ID
Mm.193924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081058.2 NP_001074527.1 2 1735
NM_027118.1 NP_081394.1 2 1735
GenBank mRNA AK035493.1 - 2 409
AK083577.1 - 2 121
AK173265.1 - 2 1735
BC076619.1 - 3 190

View Full Product Details