Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012727.1 | NP_001012745.1 | 5 | 1101 | |
NM_006412.3 | NP_006403.2 | 6 | 1197 | ||
GenBank mRNA | AF000237.1 | - | 6 | 1161 | |
AF011374.1 | - | 6 | 1158 | ||
BC000026.2 | - | 6 | 1140 | 3507150 | |
BC004529.2 | - | 5 | 1035 | 3347049 | |
BC007269.1 | - | 4 | 772 | ||
BC019292.1 | - | 6 | 1155 | 2823580 | |
U56418.2 | - | 6 | 1134 |