Gene Transcripts
- Gene Symbol
- ERLIN1
- Entrez Gene ID
- 10613
- Gene Name
- ER lipid raft associated 1
- Gene Aliases
- C10orf69, Erlin-1, KE04, KEO4, SPFH1, SPG62
- Chromosome Location
- Chr. 10: 100150090 - 100186057 on Build GRCh38
- UniGene ID
- Hs.150087
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001100626.1 |
NP_001094096.1
|
5, 6 |
410 |
|
NM_006459.3 |
NP_006450.2
|
4, 5 |
654 |
|
XM_005269442.3 |
XP_005269499.1
|
5, 6 |
440 |
|
XM_017015541.1 |
XP_016871030.1
|
5, 6 |
424 |
|
GenBank mRNA |
AF064093.1 |
- |
5, 6 |
386 |
|
AK056574.1 |
- |
4, 5 |
3011 |
|
AK222480.1 |
- |
5, 6 |
382 |
|
AK298427.1 |
- |
4, 5 |
300 |
|
AK315580.1 |
- |
5, 6 |
382 |
|
BC031791.1 |
- |
4, 5 |
323 |
4214940 |
BP280488.1 |
- |
5, 6 |
410 |
|
BU192550.1 |
- |
4, 5 |
654 |
|
View Full Product Details