Gene Transcripts
- Gene Symbol
- TOMM34
- Entrez Gene ID
- 10953
- Gene Name
- translocase of outer mitochondrial membrane 34
- Gene Aliases
- HTOM34P, TOM34, URCC3
- Chromosome Location
- Chr. 20: 44942130 - 44960520 on Build GRCh38
- UniGene ID
- Hs.517066
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006809.4 |
NP_006800.2
|
6, 7 |
973 |
|
XM_005260254.2 |
XP_005260311.1
|
6, 7 |
906 |
|
XM_011528501.1 |
XP_011526803.1
|
6, 7 |
792 |
|
GenBank mRNA |
AB085681.1 |
- |
6, 7 |
898 |
|
AK000594.1 |
- |
6, 7 |
892 |
|
AK129538.1 |
- |
5, 6 |
474 |
|
AK222952.1 |
- |
6, 7 |
880 |
|
AK302128.1 |
- |
6, 7 |
802 |
|
AK307655.1 |
- |
6, 7 |
914 |
|
BC001763.1 |
- |
6, 7 |
886 |
3544124 |
BC007423.2 |
- |
6, 7 |
886 |
2960090 |
BC014907.1 |
- |
6, 7 |
863 |
3922502 |
BT020008.1 |
- |
6, 7 |
833 |
|
BT020009.1 |
- |
6, 7 |
833 |
|
CR456765.1 |
- |
6, 7 |
833 |
|
U58970.1 |
- |
6, 7 |
864 |
|
View Full Product Details