Gene Transcripts

Gene Symbol
SLC27A3
Entrez Gene ID
11000
Gene Name
solute carrier family 27 member 3
Gene Aliases
ACSVL3, FATP3, VLCS-3
Chromosome Location
Chr. 1: 153774989 - 153780157 on Build GRCh38
UniGene ID
Hs.438723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024330.1 NP_077306.1 10 2106
GenBank mRNA AJ577572.1 - 10 2399
AK027499.1 - 10 2010
AK074134.1 - 2 4201
AK075377.1 - 10 2099
AK125102.1 - 10 3043
AY358409.1 - 10 2094
BC003041.1 - 10 2106 2822875
BC009916.2 - 10 2106 2822875
BC029792.1 - 10 1974 5195359

View Full Product Details