Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001319683.1 | NP_001306612.1 | 18 | 1798 | |
NM_053274.2 | NP_444504.1 | 19 | 1840 | ||
NR_135089.1 | 18 | 1755 | |||
XM_005270401.3 | XP_005270458.1 | 18 | 1702 | ||
XM_006710309.2 | XP_006710372.1 | 19 | 1842 | ||
XM_011540546.2 | XP_011538848.1 | 21 | 2003 | ||
XM_017000137.1 | XP_016855626.1 | 19 | 2208 | ||
XM_017000138.1 | XP_016855627.1 | 18 | 2166 | ||
XM_017000139.1 | XP_016855628.1 | 18 | 2103 | ||
XM_017000140.1 | XP_016855629.1 | 18 | 1818 | ||
XM_017000141.1 | XP_016855630.1 | 18 | 1735 | ||
XM_017000142.1 | XP_016855631.1 | 18 | 1800 | ||
XM_017000143.1 | XP_016855632.1 | 18 | 2181 | ||
XM_017000144.1 | XP_016855633.1 | 18 | 1726 | ||
GenBank mRNA | AJ302735.1 | - | 19 | 1806 | |
AJ347709.1 | - | 19 | 1807 | ||
AK295971.1 | - | 18 | 1776 | ||
BC001257.1 | - | 19 | 1807 | 3454182 | |
BG187128.1 | - | 3 | 257 | ||
U73704.1 | - | 18 | 1734 |