Gene Transcripts
- Gene Symbol
- WDR45
- Entrez Gene ID
- 11152
- Gene Name
- WD repeat domain 45
- Gene Aliases
- JM5, NBIA4, NBIA5, WDRX1, WIPI-4, WIPI4
- Chromosome Location
- Chr. X: 49074433 - 49101121 on Build GRCh38
- UniGene ID
- Hs.632807
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001029896.1 |
NP_001025067.1
|
5, 6 |
521 |
|
NM_007075.3 |
NP_009006.2
|
6, 7 |
791 |
|
GenBank mRNA |
AJ005897.1 |
- |
5, 6 |
477 |
|
AK225122.1 |
- |
5, 6 |
500 |
|
AK315717.1 |
- |
5, 6 |
508 |
|
AY691428.1 |
- |
4, 5 |
349 |
|
BC000464.2 |
- |
5, 6 |
515 |
2822742 |
BC003037.1 |
- |
5, 6 |
515 |
2822742 |
BC009027.1 |
- |
2, 3 |
145 |
|
BC069206.1 |
- |
6, 7 |
791 |
5801768 |
BQ068083.1 |
- |
5, 6 |
521 |
|
CR456725.1 |
- |
4, 5 |
349 |
|
View Full Product Details